Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304804.1 | NP_001291733.1 | 3 | 241 | ||
NM_001304805.1 | NP_001291734.1 | 4 | 390 | |||
NM_001304806.1 | NP_001291735.1 | 3 | 304 | |||
NM_173499.4 | NP_775770.1 | 3 | 459 | |||
XM_011521265.2 | XP_011519567.1 | 3 | 506 | |||
GenBank mRNA | AY489187.1 | - | 3 | 423 | ||
BC033979.2 | - | 3 | 396 | 5295377 | ||
BC066292.1 | - | 3 | 418 | 5271997 | ||
BC112390.1 | - | 2 | 787 | 5168326 | ||
HY024478.1 | - | 3 | 267 | |||
HY034703.1 | - | 4 | 387 | |||
HY047940.1 | - | 3 | 241 |