Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152455.3 | NP_689668.3 | 3 | 956 | ||
XM_006720401.3 | XP_006720464.1 | 4 | 1716 | |||
XM_011521266.2 | XP_011519568.1 | 4 | 1396 | |||
XM_017021949.1 | XP_016877438.1 | 3 | 1928 | |||
GenBank mRNA | AF525399.1 | - | 3 | 806 | ||
AK093186.1 | - | 2 | 502 | |||
AK122788.1 | - | 3 | 944 | |||
BC043612.1 | - | 3 | 924 | |||
BC062551.1 | - | 3 | 572 | |||
BC109270.1 | - | 3 | 806 | 40026738 |