Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144673.2 | NP_653274.1 | 2 | 573 | ||
GenBank mRNA | AF479260.1 | - | 2 | 565 | ||
AK098598.1 | - | 2 | 558 | |||
BC025354.1 | - | 2 | 564 | 4904062 | ||
BI830241.1 | - | 2 | 573 |