Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109763.1 | NP_001103233.1 | 4 | 664 | ||
NM_001323900.1 | NP_001310829.1 | 4 | 664 | |||
NM_001323901.1 | NP_001310830.1 | 3 | 511 | |||
NM_144675.2 | NP_653276.1 | 3 | 280 | |||
XM_011545750.2 | XP_011544052.1 | 4 | 658 | |||
XM_011545751.2 | XP_011544053.1 | 4 | 658 | |||
GenBank mRNA | AK128775.1 | - | 4 | 285 | ||
AY302134.1 | - | 2 | 114 | |||
AY358206.1 | - | 3 | 503 | |||
BC016460.1 | - | 3 | 280 | 4150450 | ||
BI752847.1 | - | 5 | 487 | |||
CV024949.1 | - | 2 | 114 |