Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172771.1 | NP_001166242.1 | 3, 4 | 711 | ||
NM_001172772.1 | NP_001166243.1 | 3, 4 | 694 | |||
NM_152456.2 | NP_689669.2 | 3, 4 | 711 | |||
XM_011522902.2 | XP_011521204.1 | 3, 4 | 795 | |||
XM_011522903.2 | XP_011521205.1 | 3, 4 | 577 | |||
GenBank mRNA | AK172744.1 | - | 2, 3 | 226 | ||
AK307727.1 | - | 2, 3 | 346 | |||
AK314913.1 | - | 2, 3 | 231 | |||
BC029804.1 | - | 2, 3 | 539 | 5192904 | ||
CD248375.1 | - | 3, 4 | 612 | |||
CD515627.1 | - | 3, 4 | 716 | |||
EU599219.1 | - | 2, 3 | 156 | |||
EU636994.1 | - | 2, 3 | 156 |