Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4RL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178568.3 | NP_848663.1 | 2 | 1570 | ||
GenBank mRNA | AF532859.1 | - | 1 | 1090 | ||
AL834409.1 | - | 1 | 1036 | |||
AY250219.1 | - | 1 | 1090 | |||
BC128608.1 | - | 2 | 1112 | 40121824 |