Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080556.1 | NP_001074025.1 | 3 | 340 | ||
NM_145054.4 | NP_659491.4 | 4 | 544 | |||
XM_017024227.1 | XP_016879716.1 | 3 | 329 | |||
GenBank mRNA | AB065281.1 | - | 4 | 509 | ||
AK074435.1 | - | 3 | 325 | |||
AK094847.1 | - | 5 | 691 | |||
AK315679.1 | - | 4 | 499 | |||
BC025392.2 | - | 4 | 504 | 4838276 | ||
DB040461.1 | - | 4 | 544 |