Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304949.1 | NP_001291878.1 | 1 | 263 | ||
NM_001304951.1 | NP_001291880.1 | 1 | 263 | |||
NM_001304953.1 | NP_001291882.1 | 1 | 263 | |||
NM_152465.2 | NP_689678.1 | 1 | 263 | |||
XM_005257927.4 | XP_005257984.1 | 1 | 415 | |||
XM_006721723.3 | XP_006721786.1 | 1 | 421 | |||
GenBank mRNA | BC029574.1 | - | 1 | 210 | 5267334 | |
BQ215318.1 | - | 1 | 103 | |||
CD300282.1 | - | 1 | 38 | |||
HY044209.1 | - | 1 | 264 |