Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139171.1 | NP_631910.1 | 3, 4 | 277 | ||
XM_011525821.1 | XP_011524123.1 | 5, 6 | 464 | |||
XM_017025554.1 | XP_016881043.1 | 3, 4 | 319 | |||
GenBank mRNA | AF480305.1 | - | 3, 4 | 277 |