Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317027.1 | NP_001303956.1 | 5 | 918 | ||
NM_001317028.1 | NP_001303957.1 | 5 | 632 | |||
NM_001317029.1 | NP_001303958.1 | 5 | 580 | |||
NM_001317030.1 | NP_001303959.1 | 2 | 485 | |||
NM_133460.2 | NP_597717.1 | 4 | 698 | |||
XM_017026304.1 | XP_016881793.1 | 5 | 935 | |||
XM_017026305.1 | XP_016881794.1 | 5 | 935 | |||
XM_017026306.1 | XP_016881795.1 | 5 | 932 | |||
XM_017026307.1 | XP_016881796.1 | 3 | 630 | |||
XM_017026308.1 | XP_016881797.1 | 3 | 630 | |||
XM_017026309.1 | XP_016881798.1 | 4 | 697 | |||
XM_017026310.1 | XP_016881799.1 | 5 | 650 | |||
XM_017026311.1 | XP_016881800.1 | 5 | 650 | |||
XM_017026312.1 | XP_016881801.1 | 4 | 698 | |||
XM_017026313.1 | XP_016881802.1 | 5 | 1673 | |||
XM_017026314.1 | XP_016881803.1 | 4 | 560 | |||
GenBank mRNA | AB075836.1 | - | 3 | 532 | ||
AK056113.1 | - | 4 | 685 | |||
AK293723.1 | - | 5 | 918 | |||
AK304632.1 | - | 2 | 479 | |||
AY695825.1 | - | 4 | 683 | |||
BC112244.1 | - | 3 | 482 | 8327712 | ||
BC113707.1 | - | 3 | 482 | 8322759 |