Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178002.2 | NP_001171473.1 | 26, 27 | 3538 | ||
NM_138813.3 | NP_620168.1 | 26, 27 | 3629 | |||
NR_047593.2 | 26, 27 | 3773 | ||||
XM_006722654.2 | XP_006722717.1 | 26, 27 | 3659 | |||
XM_006722655.2 | XP_006722718.1 | 26, 27 | 3738 | |||
XM_006722656.2 | XP_006722719.1 | 26, 27 | 3547 | |||
XM_011527707.1 | XP_011526009.1 | 26, 27 | 3632 | |||
XM_011527708.1 | XP_011526010.1 | 26, 27 | 3539 | |||
XM_011527709.1 | XP_011526011.1 | 25, 26 | 3446 | |||
XM_011527710.2 | XP_011526012.1 | 20, 21 | 2868 | |||
XM_011527712.1 | XP_011526014.1 | 11, 12 | 1661 | |||
GenBank mRNA | AK057452.1 | - | 3, 4 | 489 | ||
AK125969.1 | - | 26, 27 | 3773 | |||
AY302538.1 | - | 26, 27 | 3501 | |||
BC035162.2 | - | 26, 27 | 3624 | 5265541 |