Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145173.3 | NP_660156.1 | 2 | 616 | ||
XM_011527713.2 | XP_011526015.1 | 2 | 781 | |||
GenBank mRNA | AB076888.1 | - | 1 | 557 | ||
AY056037.1 | - | 1 | 458 | |||
AY059641.1 | - | 1 | 458 | |||
AY180973.1 | - | 1 | 458 | |||
BC030660.1 | - | 2 | 595 | 4814337 |