Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300993.2 | NP_001287922.1 | 6 | 606 | ||
NM_001321991.1 | NP_001308920.1 | 5 | 455 | |||
NM_001321992.1 | NP_001308921.1 | 4 | 786 | |||
NM_001321993.1 | NP_001308922.1 | 5 | 929 | |||
NM_001321994.1 | NP_001308923.1 | 4 | 845 | |||
NM_144694.3 | NP_653295.1 | 5 | 870 | |||
XM_011526544.2 | XP_011524846.1 | 5 | 453 | |||
XM_011526545.2 | XP_011524847.1 | 4 | 357 | |||
GenBank mRNA | AK055353.1 | - | 5 | 848 | ||
AK297561.1 | - | 4 | 547 | |||
AK308252.1 | - | 6 | 606 | |||
BC130425.1 | - | 5 | 753 | 40146456 | ||
BC130433.1 | - | 5 | 753 | 40146464 | ||
BP312352.1 | - | 5 | 455 |