Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153812.2 | NP_722519.2 | 4 | 1172 | ||
XM_011540762.1 | XP_011539064.1 | 4 | 974 | |||
GenBank mRNA | AB209544.1 | - | 4 | 864 | ||
AK027492.1 | - | 3 | 697 | |||
AK097715.1 | - | 4 | 1106 | |||
AK315110.1 | - | 4 | 1104 | |||
AL121733.1 | - | 2 | 660 | |||
AY069975.1 | - | 4 | 802 | |||
BC032792.2 | - | 2 | 599 | |||
BC038516.1 | - | 4 | 1183 | 5266750 |