Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153812.2 | NP_722519.2 | 3 | 901 | ||
XM_011540762.1 | XP_011539064.1 | 3 | 703 | |||
GenBank mRNA | AB209544.1 | - | 3 | 593 | ||
AK027492.1 | - | 2 | 426 | |||
AK097715.1 | - | 3 | 835 | |||
AK315110.1 | - | 3 | 833 | |||
AL121733.1 | - | 1 | 389 | |||
AY069975.1 | - | 3 | 531 | |||
BC032792.2 | - | 1 | 328 | |||
BC038516.1 | - | 3 | 912 | 5266750 |