Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143687.2 | NP_001137159.1 | 15 | 1821 | ||
NM_152494.3 | NP_689707.2 | 16 | 1896 | |||
GenBank mRNA | AK057347.1 | - | 16 | 1878 | ||
AK301876.1 | - | 15 | 1798 | |||
AK301902.1 | - | 15 | 1839 | |||
AK301935.1 | - | 15 | 1607 | |||
BC064844.1 | - | 15 | 1851 | 30378702 |