Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC24 SIRNA Assays ›
Gene Symbol:
CCDC24
Gene Name:
coiled-coil domain containing 24
Gene Aliases:
-
Chromosome Location:
Chr.1: 43991247 - 43996528 on Build GRCh38
UniGene ID:
Hs.632394
Species:
Human
siRNA ID
HSS135561
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152499.2
NP_689712.1
5
631
XM_017000423.1
XP_016855912.1
4
845
XM_017000424.1
XP_016855913.1
4
982
XM_017000425.1
XP_016855914.1
4
845
XM_017000426.1
XP_016855915.1
4
752
XM_017000427.1
XP_016855916.1
5
749
XM_017000428.1
XP_016855917.1
5
746
XM_017000429.1
XP_016855918.1
4
737
XM_017000430.1
XP_016855919.1
5
746
XM_017000431.1
XP_016855920.1
5
638
XM_017000432.1
XP_016855921.1
4
821
XM_017000433.1
XP_016855922.1
5
602
XM_017000434.1
XP_016855923.1
5
602
XM_017000435.1
XP_016855924.1
4
590
XM_017000436.1
XP_016855925.1
4
590
XM_017000437.1
XP_016855926.1
5
548
XM_017000438.1
XP_016855927.1
5
533
XM_017000439.1
XP_016855928.1
5
509
XM_017000440.1
XP_016855929.1
5
494
XM_017000441.1
XP_016855930.1
5
494
XM_017000442.1
XP_016855931.1
4
374
XM_017000443.1
XP_016855932.1
5
832
XM_017000445.1
XP_016855934.1
3
246
XM_017000446.1
XP_016855935.1
3
324
GenBank mRNA
AK092569.1
-
5
684
AY487420.1
-
4
352
BC033868.1
-
5
631
5207706
BC047313.1
-
4
828
BX641142.1
-
3
296
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline