Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTSF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008901.1 | NP_001008901.1 | 1 | 325 | ||
NM_176791.3 | NP_789761.1 | 1 | 325 | |||
XM_005260298.4 | XP_005260355.1 | 1 | 458 | |||
GenBank mRNA | AA813527.1 | - | 2 | 436 | ||
AK058060.1 | - | 1 | 321 | |||
BC040049.1 | - | 1 | 250 | 5744556 | ||
BQ428678.1 | - | 1 | 329 | |||
BX355734.2 | - | 1 | 245 |