Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018060.2 | NP_001018070.1 | 16 | 1649 | ||
NM_001146288.1 | NP_001139760.1 | 16 | 1734 | |||
NM_144704.2 | NP_653305.1 | 16 | 1649 | |||
NR_027464.1 | 15 | 1606 | ||||
GenBank mRNA | AK055035.1 | - | 16 | 1633 | ||
AK094844.1 | - | 16 | 1638 | |||
AK126917.1 | - | 2 | 976 | |||
AK295581.1 | - | 15 | 1509 | |||
AK316042.1 | - | 15 | 1502 | |||
AY927594.1 | - | 4 | 257 | |||
BC032485.1 | - | 16 | 1616 | 5221203 | ||
CR456342.1 | - | 15 | 1468 |