Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 7, 8 | 811 | ||
XM_011529917.2 | XP_011528219.1 | 7, 8 | 670 | |||
XM_011529918.1 | XP_011528220.1 | 7, 8 | 811 | |||
XM_017028621.1 | XP_016884110.1 | 7, 8 | 692 | |||
XM_017028622.1 | XP_016884111.1 | 7, 8 | 611 | |||
XM_017028624.1 | XP_016884113.1 | 6, 7 | 774 | |||
GenBank mRNA | AK098703.1 | - | 7, 8 | 636 | ||
BC030013.1 | - | 7, 8 | 421 | 4823215 | ||
BI459310.1 | - | 6, 7 | 379 | |||
CR456341.1 | - | 6, 7 | 398 | |||
HY002382.1 | - | 8, 9 | 461 |