Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529914.2 | XP_011528216.1 | 10 | 1078 | ||
XM_011529917.2 | XP_011528219.1 | 10 | 937 | |||
XM_011529918.1 | XP_011528220.1 | 10 | 1078 | |||
XM_017028621.1 | XP_016884110.1 | 10 | 959 | |||
XM_017028622.1 | XP_016884111.1 | 10 | 878 | |||
XM_017028624.1 | XP_016884113.1 | 9 | 1041 | |||
XM_017028625.1 | XP_016884114.1 | 5 | 429 | |||
XM_017028626.1 | XP_016884115.1 | 3 | 372 | |||
GenBank mRNA | AK098703.1 | - | 10 | 903 | ||
BC030013.1 | - | 10 | 688 | 4823215 | ||
BI459310.1 | - | 9 | 647 | |||
CR456341.1 | - | 9 | 665 |