Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026920.1 | 2 | 1176 | |||
GenBank mRNA | AK093120.1 | - | 2 | 1075 | ||
AK098685.1 | - | 1 | 557 | |||
BC042980.1 | - | 3 | 1315 | |||
BC065553.1 | - | 2 | 1069 | |||
BC117160.1 | - | 1 | 811 | |||
BC117186.1 | - | 1 | 811 |
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 26 | 4600 | ||
NM_001303257.2 | NP_001290186.1 | 26 | 4591 | |||
NM_014941.3 | NP_055756.1 | 27 | 5466 | |||
XM_017028667.1 | XP_016884156.1 | 26 | 4816 | |||
GenBank mRNA | BM683680.1 | - | 1 | 583 |