Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026920.1 | 2 | 798 | |||
GenBank mRNA | AK093120.1 | - | 2 | 697 | ||
AK098685.1 | - | 1 | 179 | |||
BC042980.1 | - | 3 | 937 | |||
BC065553.1 | - | 2 | 691 | |||
BC117160.1 | - | 1 | 433 | |||
BC117186.1 | - | 1 | 433 |
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 26 | 4978 | ||
NM_001303257.2 | NP_001290186.1 | 26 | 4969 | |||
NM_014941.3 | NP_055756.1 | 27 | 5844 | |||
XM_017028667.1 | XP_016884156.1 | 26 | 5194 | |||
GenBank mRNA | BM683680.1 | - | 1 | 205 |