Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKAP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304361.1 | NP_001291290.1 | 4 | 1345 | ||
NM_152515.4 | NP_689728.3 | 4 | 1404 | |||
NR_130712.1 | 5 | 646 | ||||
XM_011510666.2 | XP_011508968.1 | 3 | 1277 | |||
GenBank mRNA | AK097948.1 | - | 5 | 586 | ||
AK292530.1 | - | 4 | 1355 | |||
AK302875.1 | - | 4 | 1286 | |||
AK316241.1 | - | 3 | 1269 | |||
AL832036.1 | - | 4 | 1355 | |||
BC036217.1 | - | 4 | 1336 | 5270339 |