Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165977.2 | NP_001159449.1 | 8 | 1134 | ||
NM_001165978.2 | NP_001159450.1 | 8 | 1134 | |||
NM_001321070.1 | NP_001307999.1 | 8 | 1125 | |||
NM_144707.3 | NP_653308.2 | 8 | 1134 | |||
XM_006712301.2 | XP_006712364.1 | 5 | 394 | |||
XM_011510672.2 | XP_011508974.1 | 8 | 1118 | |||
GenBank mRNA | AF245303.1 | - | 8 | 1129 | ||
AF245304.1 | - | 8 | 1129 | |||
AK091175.1 | - | 8 | 1104 | |||
AK290366.1 | - | 8 | 1089 | |||
AK313267.1 | - | 8 | 1090 | |||
AY358377.1 | - | 8 | 1085 | |||
BC113877.1 | - | 6 | 885 | |||
BC114525.1 | - | 8 | 1109 | 40054105 |