Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 3, 4 | 567 | ||
NM_145280.5 | NP_660323.3 | 3, 4 | 428 | |||
XM_005246339.3 | XP_005246396.1 | 3, 4 | 623 | |||
XM_005246340.3 | XP_005246397.1 | 3, 4 | 442 | |||
XM_005246341.2 | XP_005246398.1 | 3, 4 | 549 | |||
XM_006712329.3 | XP_006712392.1 | 3, 4 | 457 | |||
XM_011510725.2 | XP_011509027.1 | 3, 4 | 406 | |||
GenBank mRNA | AF455817.1 | - | 3, 4 | 379 | ||
AK093812.1 | - | 3, 4 | 428 | |||
BC033720.1 | - | 3, 4 | 439 | 4934087 | ||
BG832122.1 | - | 3, 4 | 485 |