Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199148.1 | NP_001186077.1 | 10 | 1243 | ||
NM_173512.2 | NP_775783.1 | 9 | 1177 | |||
XM_005246350.4 | XP_005246407.1 | 11 | 1396 | |||
XM_006712337.3 | XP_006712400.1 | 10 | 1392 | |||
XM_011510737.2 | XP_011509039.1 | 10 | 1488 | |||
XM_011510742.2 | XP_011509044.1 | 8 | 1184 | |||
XM_017003455.1 | XP_016858944.1 | 11 | 1632 | |||
XM_017003456.1 | XP_016858945.1 | 10 | 1566 | |||
XM_017003457.1 | XP_016858946.1 | 10 | 1557 | |||
XM_017003458.1 | XP_016858947.1 | 10 | 1517 | |||
XM_017003459.1 | XP_016858948.1 | 9 | 1255 | |||
XM_017003460.1 | XP_016858949.1 | 11 | 1632 | |||
XM_017003461.1 | XP_016858950.1 | 11 | 1632 | |||
XM_017003462.1 | XP_016858951.1 | 11 | 1632 | |||
XM_017003463.1 | XP_016858952.1 | 10 | 1029 | |||
XM_017003464.1 | XP_016858953.1 | 7 | 1241 | |||
XM_017003465.1 | XP_016858954.1 | 6 | 769 | |||
GenBank mRNA | AK097141.1 | - | 9 | 1177 | ||
AK293998.1 | - | 10 | 1204 | |||
BC125157.1 | - | 12 | 1170 | 40120878 |