Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199148.1 | NP_001186077.1 | 5, 6 | 710 | ||
NM_173512.2 | NP_775783.1 | 4, 5 | 644 | |||
XM_005246350.4 | XP_005246407.1 | 6, 7 | 863 | |||
XM_006712337.3 | XP_006712400.1 | 6, 7 | 1021 | |||
XM_011510737.2 | XP_011509039.1 | 5, 6 | 955 | |||
XM_017003455.1 | XP_016858944.1 | 6, 7 | 1021 | |||
XM_017003456.1 | XP_016858945.1 | 5, 6 | 955 | |||
XM_017003457.1 | XP_016858946.1 | 5, 6 | 946 | |||
XM_017003458.1 | XP_016858947.1 | 5, 6 | 906 | |||
XM_017003459.1 | XP_016858948.1 | 4, 5 | 644 | |||
XM_017003460.1 | XP_016858949.1 | 6, 7 | 1021 | |||
XM_017003461.1 | XP_016858950.1 | 6, 7 | 1021 | |||
XM_017003462.1 | XP_016858951.1 | 6, 7 | 1021 | |||
XM_017003463.1 | XP_016858952.1 | 5, 6 | 496 | |||
XM_017003464.1 | XP_016858953.1 | 2, 3 | 630 | |||
GenBank mRNA | AK097141.1 | - | 4, 5 | 644 | ||
AK293998.1 | - | 5, 6 | 671 | |||
BC125157.1 | - | 7, 8 | 637 | 40120878 |