Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDSUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128212.2 | NP_001121684.1 | 9 | 1266 | ||
NM_001128213.1 | NP_001121685.1 | 9 | 1282 | |||
NM_001307994.1 | NP_001294923.1 | 5 | 990 | |||
NM_152528.2 | NP_689741.2 | 9 | 1174 | |||
XM_005246355.2 | XP_005246412.2 | 9 | 1260 | |||
XM_006712341.3 | XP_006712404.1 | 8 | 754 | |||
XM_011510753.2 | XP_011509055.1 | 9 | 1226 | |||
XM_011510754.2 | XP_011509056.1 | 9 | 960 | |||
XM_017003481.1 | XP_016858970.1 | 9 | 1260 | |||
XM_017003482.1 | XP_016858971.1 | 4 | 891 | |||
XM_017003483.1 | XP_016858972.1 | 8 | 1081 | |||
GenBank mRNA | AK093494.1 | - | 9 | 1174 | ||
AK129983.1 | - | 9 | 1282 | |||
AL040518.1 | - | 8 | 650 | |||
BC029520.1 | - | 5 | 894 | 5301559 |