Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042601.2 | NP_001036066.1 | 10 | 3810 | ||
NM_133462.3 | NP_597719.1 | 12 | 2056 | |||
XM_011512439.2 | XP_011510741.1 | 8 | 1298 | |||
XM_017005739.1 | XP_016861228.1 | 11 | 1887 | |||
XM_017005740.1 | XP_016861229.1 | 11 | 2845 | |||
GenBank mRNA | AB075860.2 | - | 10 | 1594 | ||
AK055984.1 | - | 12 | 2014 | |||
AL832052.1 | - | 10 | 3749 | |||
AL832310.1 | - | 1 | 420 | |||
AL833086.2 | - | 10 | 3746 | |||
BC027990.1 | - | 12 | 1989 | 4998121 | ||
BX648669.1 | - | 5 | 6231 |