Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGO1 SIRNA Assays ›
Gene Symbol:
SGO1
Gene Name:
shugoshin 1
Gene Aliases:
CAID, NY-BR-85, SGO, SGOL1
Chromosome Location:
Chr.3: 20160593 - 20188143 on Build GRCh38
UniGene ID:
Hs.105153
Species:
Human
siRNA ID
HSS135742
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012409.3
NP_001012409.1
2, 3
319
NM_001012410.4
NP_001012410.1
2, 3
319
NM_001012411.3
NP_001012411.1
2, 3
319
NM_001012412.4
NP_001012412.1
2, 3
319
NM_001012413.3
NP_001012413.1
2, 3
319
NM_001199251.2
NP_001186180.1
2, 3
430
NM_001199252.2
NP_001186181.1
2, 3
430
NM_001199253.2
NP_001186182.1
2, 3
430
NM_001199254.2
NP_001186183.1
2, 3
430
NM_001199255.2
NP_001186184.1
2, 3
430
NM_001199256.2
NP_001186185.1
2, 3
430
NM_001199257.2
NP_001186186.1
2, 3
430
NM_138484.4
NP_612493.1
2, 3
319
XM_011533373.2
XP_011531675.1
2, 3
403
XM_011533375.2
XP_011531677.1
2, 3
772
XM_011533376.2
XP_011531678.1
2, 3
627
XM_011533377.2
XP_011531679.1
2, 3
1607
GenBank mRNA
AB187577.1
-
2, 3
277
AB187578.1
-
2, 3
277
AB187579.1
-
2, 3
277
AB187580.1
-
2, 3
277
AB187581.1
-
2, 3
277
AB187582.1
-
2, 3
277
AB190994.1
-
2, 3
167
AB193056.1
-
2, 3
324
AB193057.1
-
2, 3
213
AB193058.1
-
2, 3
213
AB193059.1
-
2, 3
324
AB193060.1
-
2, 3
324
AB193061.1
-
2, 3
213
AB193062.1
-
2, 3
324
AB193063.1
-
2, 3
213
AB193064.1
-
2, 3
324
AB193065.1
-
2, 3
324
AB193066.1
-
2, 3
213
AB451340.1
-
1, 2
137
AB451489.1
-
1, 2
137
AK308095.1
-
2, 3
418
BC001339.2
-
1, 2
78
BC017867.1
-
2, 3
277
4685140
BG530826.1
-
2, 3
291
BX494477.1
-
2, 3
433
BX648516.1
-
2, 3
404
DB163747.1
-
2, 3
429
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
attachment of spindle microtubules to kinetochore
centriole-centriole cohesion
meiotic chromosome segregation
cell division
protein binding
kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline