Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199511.2 | NP_955805.1 | 2 | 1631 | ||
NM_199512.2 | NP_955806.1 | 2 | 1034 | |||
GenBank mRNA | AB052098.1 | - | 2 | 1248 | ||
AB266387.1 | - | 2 | 1819 | |||
AF506819.1 | - | 2 | 948 | |||
AK075210.1 | - | 1 | 869 | |||
AK292881.1 | - | 2 | 1315 | |||
AL833034.1 | - | 2 | 902 | |||
AY333429.1 | - | 2 | 1528 | |||
AY526612.1 | - | 2 | 1619 | |||
AY548106.1 | - | 2 | 1604 | |||
AY548107.1 | - | 2 | 1604 | |||
AY726593.1 | - | 2 | 1085 | |||
BC042128.1 | - | 1 | 562 | |||
BC086876.1 | - | 2 | 1189 | |||
BC110842.1 | - | 2 | 1616 | 6205672 | ||
BC116178.1 | - | 2 | 1521 | 40068210 | ||
BX647117.1 | - | 2 | 1505 | |||
BX647817.1 | - | 2 | 920 |