Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199511.2 | NP_955805.1 | 8 | 3627 | ||
NM_199512.2 | NP_955806.1 | 8 | 3030 | |||
GenBank mRNA | AB052098.1 | - | 8 | 3244 | ||
AB266387.1 | - | 8 | 3815 | |||
AF506819.1 | - | 8 | 2944 | |||
AK292881.1 | - | 8 | 3311 | |||
AK304561.1 | - | 8 | 1833 | |||
AL833034.1 | - | 8 | 2898 | |||
AY333429.1 | - | 8 | 3524 | |||
AY526612.1 | - | 8 | 3615 | |||
AY548106.1 | - | 8 | 3600 | |||
AY548107.1 | - | 8 | 3600 | |||
AY726593.1 | - | 8 | 3080 | |||
BC042105.1 | - | 4 | 564 | |||
BC073164.1 | - | 7 | 942 | |||
BC110842.1 | - | 8 | 3612 | 6205672 | ||
BC116178.1 | - | 8 | 3517 | 40068210 | ||
BX647117.1 | - | 8 | 3501 | |||
BX647817.1 | - | 8 | 2916 |