Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287010.1 | NP_001273939.1 | 2, 3 | 355 | ||
NM_001287011.1 | NP_001273940.1 | 2, 3 | 355 | |||
NM_001287013.1 | NP_001273942.1 | 2, 3 | 192 | |||
NM_022343.3 | NP_071738.1 | 2, 3 | 355 | |||
NR_104637.1 | 4, 5 | 736 | ||||
NR_104638.1 | 2, 3 | 147 | ||||
NR_104639.1 | 2, 3 | 355 | ||||
GenBank mRNA | AJ011129.1 | - | 1, 2 | 112 | ||
AK074488.1 | - | 2, 3 | 177 | |||
AK075091.1 | - | 2, 3 | 156 | |||
AK091288.1 | - | 3, 4 | 969 | |||
AK298741.1 | - | 2, 3 | 156 | |||
AK307243.1 | - | 2, 3 | 156 | |||
AY039756.1 | - | 2, 3 | 146 | |||
BC017918.1 | - | 2, 3 | 158 | 4296721 | ||
BF515272.1 | - | 2, 3 | 355 | |||
CR749781.1 | - | 4, 5 | 537 | |||
DA268281.1 | - | 2, 3 | 192 | |||
DA271525.1 | - | 2, 3 | 132 | |||
DA986727.1 | - | 2, 3 | 156 |