Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287013.1 | NP_001273942.1 | 4, 5 | 390 | ||
NM_022343.3 | NP_071738.1 | 4, 5 | 553 | |||
NR_104637.1 | 6, 7 | 934 | ||||
NR_104638.1 | 4, 5 | 345 | ||||
NR_104640.1 | 4, 5 | 575 | ||||
GenBank mRNA | AJ011129.1 | - | 3, 4 | 310 | ||
AK075091.1 | - | 4, 5 | 354 | |||
AK091288.1 | - | 5, 6 | 1167 | |||
AY039756.1 | - | 4, 5 | 344 | |||
CR749781.1 | - | 6, 7 | 735 | |||
DA268281.1 | - | 4, 5 | 390 | |||
DA271525.1 | - | 4, 5 | 330 | |||
DA824243.1 | - | 4, 5 | 344 |