Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182523.1 | NP_872329.1 | 4 | 468 | ||
XM_011533402.2 | XP_011531704.1 | 6 | 645 | |||
XM_017005760.1 | XP_016861249.1 | 5 | 427 | |||
GenBank mRNA | AF086095.1 | - | 2 | 630 | ||
BC052644.1 | - | 4 | 468 | 6163781 | ||
BX648671.1 | - | 4 | 495 | |||
CR749370.1 | - | 3 | 3603 |