Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144718.3 | NP_653319.1 | 13 | 1913 | ||
XM_011512461.2 | XP_011510763.1 | 13 | 1670 | |||
XM_017005775.1 | XP_016861264.1 | 13 | 1670 | |||
XM_017005776.1 | XP_016861265.1 | 13 | 1807 | |||
XM_017005777.1 | XP_016861266.1 | 13 | 1730 | |||
XM_017005778.1 | XP_016861267.1 | 13 | 1914 | |||
GenBank mRNA | AK126897.1 | - | 9 | 1319 | ||
AK129575.1 | - | 3 | 442 | |||
AL832148.1 | - | 13 | 1882 | |||
AL832327.2 | - | 13 | 3578 | |||
AY099107.1 | - | 13 | 1833 | |||
BC036951.1 | - | 13 | 1854 | 5500345 | ||
BC038595.1 | - | 1 | 36 |