Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152539.2 | NP_689752.2 | 1 | 1253 | ||
GenBank mRNA | AK057421.1 | - | 1 | 1253 | ||
BC130475.1 | - | 1 | 1252 | 40146506 | ||
BC144197.1 | - | 1 | 1252 | 9052717 | ||
DW009752.1 | - | 1 | 28 |