Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152540.3 | NP_689753.2 | 8, 9 | 2086 | ||
GenBank mRNA | AB074276.1 | - | 8, 9 | 2059 | ||
AK024713.1 | - | 2, 3 | 223 | |||
AK096833.1 | - | 7, 8 | 1937 | |||
AY299407.1 | - | 8, 9 | 2086 | |||
BC012458.1 | - | 8, 9 | 1852 | |||
BC021137.1 | - | 8, 9 | 2043 | 4866354 | ||
BC032453.1 | - | 8, 9 | 2084 | 5176029 |