Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286052.1 | NP_001272981.1 | 3 | 986 | ||
NM_001286053.1 | NP_001272982.1 | 2 | 859 | |||
NM_001286054.1 | NP_001272983.1 | 5 | 1183 | |||
NM_182524.3 | NP_872330.1 | 4 | 1082 | |||
XM_011513398.2 | XP_011511700.1 | 4 | 1701 | |||
GenBank mRNA | AK056302.1 | - | 4 | 1036 | ||
AK300505.1 | - | 3 | 976 | |||
AK301848.1 | - | 2 | 825 | |||
BC036110.1 | - | 4 | 1046 | 5297405 | ||
BC043151.1 | - | 4 | 1051 | 5297425 | ||
BX537887.1 | - | 4 | 1034 |