Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300735.1 | NP_001287664.1 | 3 | 401 | ||
NM_152545.2 | NP_689758.1 | 3 | 401 | |||
XM_017007813.1 | XP_016863302.1 | 3 | 489 | |||
GenBank mRNA | AK056257.1 | - | 3 | 380 | ||
AK291459.1 | - | 3 | 349 | |||
BC036784.1 | - | 3 | 363 | |||
BC121003.1 | - | 3 | 316 | 40116919 | ||
BX648337.1 | - | 3 | 314 |