Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC38A9 SIRNA Assays ›
Gene Symbol:
SLC38A9
Gene Name:
solute carrier family 38 member 9
Gene Aliases:
URLC11
Chromosome Location:
Chr.5: 55625845 - 55712344 on Build GRCh38
UniGene ID:
Hs.742101
Species:
Human
siRNA ID
HSS135864
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC38A9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258286.1
NP_001245215.1
4
746
NM_001258287.1
NP_001245216.1
5
850
NM_001282429.1
NP_001269358.1
5
463
NM_173514.3
NP_775785.2
6
596
NR_047649.1
4
746
XM_006714537.2
XP_006714600.1
8
830
XM_006714538.3
XP_006714601.1
7
717
XM_006714539.3
XP_006714602.1
7
760
XM_011543173.1
XP_011541475.1
8
818
XM_011543174.1
XP_011541476.1
9
916
XM_011543176.1
XP_011541478.1
8
830
XM_011543177.1
XP_011541479.1
8
830
XM_011543178.1
XP_011541480.1
8
830
XM_011543180.1
XP_011541482.1
8
830
XM_011543182.1
XP_011541484.1
8
830
XM_011543183.1
XP_011541485.1
8
830
XM_011543184.1
XP_011541486.1
8
830
XM_017009077.1
XP_016864566.1
6
528
XM_017009078.1
XP_016864567.1
6
605
XM_017009079.1
XP_016864568.1
6
528
XM_017009080.1
XP_016864569.1
7
717
XM_017009081.1
XP_016864570.1
6
528
XM_017009082.1
XP_016864571.1
6
528
GenBank mRNA
AB105188.1
-
6
528
AK075190.1
-
6
528
AK122957.1
-
5
443
AK127988.1
-
5
490
AK301850.1
-
4
408
BC066891.1
-
6
585
4373965
BC101362.2
-
6
527
40025719
Back To Top
More Information
Panther Classification:
Molecular Function -
amino acid transporter
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transmembrane transport
amino acid transport
positive regulation of TOR signaling
cellular response to amino acid stimulus
protein binding
amino acid transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-tl62x:80/100.66.78.111:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline