Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181776.2 | NP_861441.2 | 4, 5 | 580 | ||
XM_005268377.3 | XP_005268434.1 | 4, 5 | 575 | |||
XM_006714756.3 | XP_006714819.1 | 4, 5 | 575 | |||
XM_017009083.1 | XP_016864572.1 | 4, 5 | 575 | |||
GenBank mRNA | AK297682.1 | - | 4, 5 | 546 | ||
AK309575.1 | - | 4, 5 | 580 | |||
AY162214.1 | - | 4, 5 | 548 | |||
BC101101.1 | - | 4, 5 | 518 | 40010274 | ||
BC101103.2 | - | 4, 5 | 518 | 40010278 |