Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 10 | 1562 | ||
NM_153223.3 | NP_694955.2 | 11 | 1643 | |||
XM_005271901.4 | XP_005271958.1 | 10 | 1424 | |||
XM_011543185.2 | XP_011541487.1 | 10 | 1977 | |||
XM_011543186.2 | XP_011541488.1 | 7 | 1256 | |||
XM_017009085.1 | XP_016864574.1 | 7 | 1121 | |||
GenBank mRNA | AK093409.1 | - | 3 | 329 | ||
AK095646.1 | - | 10 | 1564 | |||
AK296999.1 | - | 11 | 1607 | |||
BC040527.1 | - | 10 | 2226 | |||
BC144558.1 | - | 13 | 1770 | |||
BX648687.1 | - | 1 | 36 | |||
CR627324.1 | - | 6 | 2799 |