Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 13 | 2024 | ||
NM_153223.3 | NP_694955.2 | 14 | 2105 | |||
XM_005271901.4 | XP_005271958.1 | 13 | 1886 | |||
XM_011543185.2 | XP_011541487.1 | 13 | 2439 | |||
XM_011543186.2 | XP_011541488.1 | 10 | 1718 | |||
XM_017009085.1 | XP_016864574.1 | 10 | 1583 | |||
GenBank mRNA | AK093409.1 | - | 6 | 791 | ||
AK095646.1 | - | 13 | 2026 | |||
AL833929.1 | - | 2 | 212 | |||
BC040527.1 | - | 13 | 2688 | |||
BC144558.1 | - | 16 | 2232 | |||
BX648687.1 | - | 4 | 498 |