Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017373.3 | NP_001017373.2 | 4 | 490 | ||
NM_001258275.2 | NP_001245204.1 | 6 | 771 | |||
NM_001277185.1 | NP_001264114.1 | 3 | 272 | |||
XM_017010305.1 | XP_016865794.1 | 3 | 320 | |||
XM_017010306.1 | XP_016865795.1 | 2 | 530 | |||
XM_017010307.1 | XP_016865796.1 | 2 | 530 | |||
XM_017010308.1 | XP_016865797.1 | 2 | 183 | |||
XM_017010309.1 | XP_016865798.1 | 2 | 183 | |||
XM_017010310.1 | XP_016865799.1 | 2 | 183 | |||
GenBank mRNA | AK091351.1 | - | 6 | 773 | ||
AK091882.1 | - | 4 | 490 | |||
AK302228.1 | - | 3 | 274 | |||
AK310075.1 | - | 2 | 526 | |||
BC029851.1 | - | 2 | 179 | |||
BC119757.1 | - | 2 | 179 | |||
BC119758.1 | - | 2 | 179 | |||
BC127653.1 | - | 2 | 177 | |||
BC127654.1 | - | 2 | 177 | |||
DA584636.1 | - | 2 | 462 |