Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017373.3 | NP_001017373.2 | 6 | 869 | ||
NM_001258275.2 | NP_001245204.1 | 8 | 1150 | |||
NM_001277185.1 | NP_001264114.1 | 5 | 651 | |||
XM_017010305.1 | XP_016865794.1 | 5 | 696 | |||
XM_017010306.1 | XP_016865795.1 | 4 | 909 | |||
XM_017010307.1 | XP_016865796.1 | 4 | 906 | |||
XM_017010308.1 | XP_016865797.1 | 4 | 562 | |||
XM_017010309.1 | XP_016865798.1 | 4 | 562 | |||
XM_017010310.1 | XP_016865799.1 | 4 | 562 | |||
GenBank mRNA | AK091351.1 | - | 8 | 1152 | ||
AK091882.1 | - | 6 | 869 | |||
AK302228.1 | - | 5 | 653 | |||
AK310075.1 | - | 4 | 905 | |||
BC029851.1 | - | 4 | 558 | |||
BC119758.1 | - | 4 | 558 | |||
BC127653.1 | - | 4 | 556 | |||
BC127654.1 | - | 4 | 556 |