Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271862.1 | NP_001258791.1 | 5 | 326 | ||
NM_173516.2 | NP_775787.1 | 5 | 425 | |||
XM_011535491.2 | XP_011533793.1 | 5 | 352 | |||
XM_017010312.1 | XP_016865801.1 | 5 | 1114 | |||
XM_017010313.1 | XP_016865802.1 | 3 | 1000 | |||
XM_017010314.1 | XP_016865803.1 | 5 | 1114 | |||
XM_017010315.1 | XP_016865804.1 | 3 | 204 | |||
XM_017010316.1 | XP_016865805.1 | 3 | 269 | |||
GenBank mRNA | AK097559.1 | - | 5 | 405 |