Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286398.2 | NP_001273327.1 | 3 | 1136 | ||
NM_152553.4 | NP_689766.1 | 5 | 602 | |||
NR_104440.2 | 4 | 493 | ||||
NR_136734.1 | 4 | 1257 | ||||
XM_011535494.2 | XP_011533796.1 | 3 | 1243 | |||
XM_011535496.2 | XP_011533798.1 | 2 | 1017 | |||
XM_011535500.2 | XP_011533802.1 | 3 | 371 | |||
GenBank mRNA | AK093417.1 | - | 4 | 493 | ||
AK122811.1 | - | 3 | 965 | |||
BC026087.2 | - | 5 | 598 | 4827114 | ||
CO402490.1 | - | 576 | ||||
DB238637.1 | - | 4 | 470 |