Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 5 | 842 | ||
NM_001300737.1 | NP_001287666.1 | 6 | 906 | |||
NM_001300738.1 | NP_001287667.1 | 6 | 856 | |||
NM_001300740.1 | NP_001287669.1 | 5 | 480 | |||
NM_153355.4 | NP_699186.2 | 4 | 641 | |||
XM_011535501.2 | XP_011533803.1 | 5 | 730 | |||
XM_011535502.2 | XP_011533804.1 | 5 | 497 | |||
XM_011535503.2 | XP_011533805.1 | 4 | 767 | |||
XM_017010318.1 | XP_016865807.1 | 4 | 765 | |||
XM_017010319.1 | XP_016865808.1 | 4 | 529 | |||
GenBank mRNA | AK055326.1 | - | 5 | 475 | ||
AK308927.1 | - | 6 | 494 | |||
AK309385.1 | - | 6 | 455 | |||
AK309407.1 | - | 2 | 217 | |||
AK309802.1 | - | 6 | 494 | |||
AK310763.1 | - | 4 | 316 | |||
AK315886.1 | - | 5 | 481 | |||
BC035062.2 | - | 6 | 608 | 5264304 | ||
DB174077.1 | - | 3 | 316 |